missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylurethane 98.0+%, TCI America™

Supplier:  TCI America P016525G

 View more versions of this product

Catalog No. P016525G

Explore available promotions

Description

Description

Specifications

Specifications

Phenylurethane
52°C
237°C
MFCD00026806
n-phenylurethane, phenylurethane, ethyl phenylcarbamate, phenylurethan, ethyl carbanilate, euphorin, keimstop, urethane, phenyl, carbanilic acid, ethyl ester, phenylethyl carbamate
CCOC(=O)NC1=CC=CC=C1
165.192
165.19
Crystalline Powder
101-99-5
White-Yellow
C9H11NO2
25 g
LBKPGNUOUPTQKA-UHFFFAOYSA-N
ethyl N-phenylcarbamate
7591
≥98.0% (GC)

Chemical Identifiers

101-99-5
165.192
LBKPGNUOUPTQKA-UHFFFAOYSA-N
7591
CCOC(=O)NC1=CC=CC=C1
C9H11NO2
MFCD00026806
n-phenylurethane, phenylurethane, ethyl phenylcarbamate, phenylurethan, ethyl carbanilate, euphorin, keimstop, urethane, phenyl, carbanilic acid, ethyl ester, phenylethyl carbamate
ethyl N-phenylcarbamate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : FD8925000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions