missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Blood Collection Tray

Designed with phlebotomists in mind. Fisherbrand™ Blood Collection Tray was developed after extensive research in clinics and hospitals. This lightweight solution for organizing and transporting tubes and smaller items includes a sturdy plastic tray, 36-well tube rack and disposable inserts.

$139.81 - $139.81

Specifications

Color White
Material Polystyrene
Product Type Phlebotomy Tray
For Use With (Application) Blood Collection
Includes Extra-deep compartment
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
22-267-272
View Documents
N/A 13 mm Tube Rack and Blood Collection Tray (Style A)
Pack of 4 Each for $139.81
 
22-267-273
View Documents
N/A 16 mm Tube Rack and Blood Collection Tray (Style B)
Pack of 4 Each for $139.81
 
Description

Description

  • Available for a 13 or 16 mm tube rack
  • Insert is removable, washable, and disposable
  • Includes an extra-deep compartment for sharps container
  • Inserts sold in packs of four (4)
Specifications

Specifications

White
Phlebotomy Tray
Extra-deep compartment
Polystyrene
Blood Collection
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'