missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phloretin, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 307651000

 View more versions of this product

Catalog No. AC307651000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phloretin
260.0°C to 262.0°C
Authentic
Glass bottle
1.5730 to 1.5750
MFCD00002288
15, 7437
Solubility in water: .. Other solubilities: soluble in methanol
C1=CC(=CC=C1CCC(=O)C2=C(C=C(C=C2O)O)O)O
274.27
CHEBI:17276
98%
60-82-2
White to Beige
97.5% min. (HPLC)
C15H14O5
HOC6H4CH2COC6H2(OH)3
100 mg
phloretin, dihydronaringenin, 3-4-hydroxyphenyl-1-2,4,6-trihydroxyphenyl propan-1-one, phloretol, 3-4-hydroxyphenyl-1-2,4,6-trihydroxyphenyl-1-propanone, unii-s5j5oe47mk, 1-propanone, 3-4-hydroxyphenyl-1-2,4,6-trihydroxyphenyl, ccris 7459, beta-p-hydroxyphenyl phloropropiophenone, 2',4',6'-trihydroxy-3-p-hydroxyphenyl propiophenone
VGEREEWJJVICBM-UHFFFAOYSA-N
3-(4-hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)propan-1-one
4788
274.27
Powder

Chemical Identifiers

60-82-2
274.27
VGEREEWJJVICBM-UHFFFAOYSA-N
4788
3-(4-hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)propan-1-one
C15H14O5
MFCD00002288
phloretin, dihydronaringenin, 3-4-hydroxyphenyl-1-2,4,6-trihydroxyphenyl propan-1-one, phloretol, 3-4-hydroxyphenyl-1-2,4,6-trihydroxyphenyl-1-propanone, unii-s5j5oe47mk, 1-propanone, 3-4-hydroxyphenyl-1-2,4,6-trihydroxyphenyl, ccris 7459, beta-p-hydroxyphenyl phloropropiophenone, 2',4',6'-trihydroxy-3-p-hydroxyphenyl propiophenone
CHEBI:17276
C1=CC(=CC=C1CCC(=O)C2=C(C=C(C=C2O)O)O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 200-488-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only