missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosgene iminium chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 172210250

 View more versions of this product

Catalog No. AC172210250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phosgene iminium chloride
33842-02-3
Yellow
97.5% min. (Argentometry)
C3H6Cl3N
MFCD00011870
04,135; 05,195; 06,170; 08,156
dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride
NRNFKRFWZQQDMD-UHFFFAOYSA-M
dichloromethylidene(dimethyl)azanium;chloride
2723945
98%
98%
181°C to 185°C
Authentic
Glass bottle
Cl2C=N(CH3)2Cl
25 g
15, 10174
Solubility in water: hydrolysis
[Cl-].CN(C)[C+](Cl)Cl
162.44
162.45
Crystalline Powder

Chemical Identifiers

33842-02-3
162.44
NRNFKRFWZQQDMD-UHFFFAOYSA-M
2723945
C3H6Cl3N
MFCD00011870
dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride
[Cl-].CN(C)[C+](Cl)Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 251-695-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only