missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphate, Ion chromatography standard solution, Specpure™ PO4∣-^3 1000μg/mL, Thermo Scientific Chemicals

$241.05 - $438.29

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
MDL Number MFCD00003527,MFCD00146206
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
Synonym Phosphate, Ion chromatography standard solution
PubChem CID 23672064
ChEBI CHEBI:37585
IUPAC Name sodium;dihydrogen phosphate
SMILES [Na+].OP(O)([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA35548AE
View Documents
Thermo Scientific Chemicals
035548AE
100 mL
Each for $241.05
 
AA35548AP
View Documents
Thermo Scientific Chemicals
035548AP
500 mL
Each for $438.29
 
Description

Description

Phosphate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is used for calibration of ion chromatography and other analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7558-80-7
119.98
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
23672064
sodium;dihydrogen phosphate
H2NaO4P
MFCD00003527,MFCD00146206
Phosphate, Ion chromatography standard solution
CHEBI:37585
[Na+].OP(O)([O-])=O

Specifications

PO4-3 1000μg/mL
100 mL
MFCD00003527,MFCD00146206
Miscible with water
Liquid
sodium;dihydrogen phosphate
23672064
Phosphate
7558-80-7
H2NaO4P
Phosphate, Ion chromatography standard solution
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
[Na+].OP(O)([O-])=O
119.98
CHEBI:37585
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only