missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phosphoenolpyruvic acid mono(cyclohexylammonium) salt, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1238506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phosphoenolpyruvic acid mono (cyclohexylammonium) salt | |
∼140°C (decomposition) | |
MFCD00036375 | |
cyclohexanamine 2-phosphonooxy acrylate, phosphoenolpyruvic acid cyclohexylammonium salt, 2-propenoic acid, 2-phosphonooxy-, compd. with cyclohexanamine 1:1, cyclohexylamine; phosphoenolpyruvic acid, phosphoenolpyruvic acid cyclohexylamine salt, phosphoenolpyruvic acid, cyclohexylammonium salt, cyclohexanamine; 2-phosphonooxyprop-2-enoic acid, phospho enol pyruvic acid cyclohexylammonium salt, phosphoenolpyruvic acid monocyclohexylammonium salt | |
NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O | |
267.22 | |
267.22 |
10526-80-4 | |
C9H18NO6P | |
5 g | |
VHFCNZDHPABZJO-UHFFFAOYSA-N | |
cyclohexanamine;2-phosphonooxyprop-2-enoic acid | |
82702 | |
98% |
Chemical Identifiers
10526-80-4 | |
267.22 | |
VHFCNZDHPABZJO-UHFFFAOYSA-N | |
82702 | |
NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O |
C9H18NO6P | |
MFCD00036375 | |
cyclohexanamine 2-phosphonooxy acrylate, phosphoenolpyruvic acid cyclohexylammonium salt, 2-propenoic acid, 2-phosphonooxy-, compd. with cyclohexanamine 1:1, cyclohexylamine; phosphoenolpyruvic acid, phosphoenolpyruvic acid cyclohexylamine salt, phosphoenolpyruvic acid, cyclohexylammonium salt, cyclohexanamine; 2-phosphonooxyprop-2-enoic acid, phospho enol pyruvic acid cyclohexylammonium salt, phosphoenolpyruvic acid monocyclohexylammonium salt | |
cyclohexanamine;2-phosphonooxyprop-2-enoic acid |
Safety and Handling
EINECSNumber : 234-084-7
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only