missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Phosphoric Acid, NF, 85-88%, Spectrum™ Chemical
PH175, 7664-38-2, H3 PO4
Chemical Identifiers
CAS
7664-38-2
Molecular Formula
H3O4P
Molecular Weight (g/mol)
97.99
InChI Key
NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name
phosphoric acid
SMILES
OP(O)(O)=O
View More Specs
Products
2
Description
Description
Spectrum™ Chemical Phosphoric Acid, NF - All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Chemical Identifiers
CAS
7664-38-2
Molecular Weight (g/mol)
97.99
IUPAC Name
phosphoric acid
Molecular Formula
H3O4P
InChI Key
NBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILES
OP(O)(O)=O
Specifications
CAS
7664-38-2
Packaging
Amber Glass Bottle
Molecular Formula
H3O4P
SMILES
OP(O)(O)=O
Molecular Weight (g/mol)
97.99
Grade
NF
CAS Max %
1
Quantity
100 mL
InChI Key
NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name
phosphoric acid
Percent Purity
85 to 88%
SDS
missing translation for 'documents'