missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, NF, 85-88%, Spectrum™ Chemical

PH175, 7664-38-2, H3PO4

$608.63 - $6,153.97

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O
View More Specs

Products 11
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18605178
View Documents
Spectrum Chemical Mfg Cor
PH175500MLGL
500 mL Amber Glass Bottle N/A
Only null left
 
18605182
View Documents
Spectrum Chemical Mfg Cor
PH17525LTGL
2.5 L Glass Bottle N/A
Only null left
 
18605181
View Documents
Spectrum Chemical Mfg Cor
PH17525LTPV
2.5 L Poly-Coated Glass Bottle N/A
Only null left
 
18605179
View Documents
Spectrum Chemical Mfg Cor
PH175500MLGLCS6
6 x 500 mL Amber Glass Bottle N/A
Only null left
 
18605183
View Documents
Spectrum Chemical Mfg Cor
PH1754LTPV
4 L Poly-Coated Glass Bottle N/A
Only null left
 
18605184
View Documents
Spectrum Chemical Mfg Cor
PH1751LTCS6
6 x 1 L Amber Glass Bottle N/A
Only null left
 
18605185
View Documents
Spectrum Chemical Mfg Cor
PH17525LTGLCS4
4 x 2.5 L Glass Bottle N/A
Only null left
 
18605186
View Documents
Spectrum Chemical Mfg Cor
PH17525LTPVCS4
4 x 2.5 L Poly-Coated Glass Bottle N/A
Only null left
 
18605187
View Documents
Spectrum Chemical Mfg Cor
PH1754LTPVCS4
4 x 4 L Poly-Coated Glass Bottle N/A
Only null left
 
18605188
View Documents
Spectrum Chemical Mfg Cor
PH17520LTBL
20 L Poly Drum N/A
Only null left
 
18605189
View Documents
Spectrum Chemical Mfg Cor
PH175200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Phosphoric Acid, NF - All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

7664-38-2
97.99
phosphoric acid
H3O4P
NBIIXXVUZAFLBC-UHFFFAOYSA-N
OP(O)(O)=O

Specifications

7664-38-2,7732-18-5
Amber Glass Bottle
H3O4P
OP(O)(O)=O
97.99
NF
85 to 88%,12 to 15%
500 mL
NBIIXXVUZAFLBC-UHFFFAOYSA-N
phosphoric acid
85 to 88%
Phosphoric Acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'