missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phosphoric acid, TraceSELECT™, Honeywell Fluka™
TraceSELECT™, for trace analysis, ∼85%
Supplier: Honeywell Chemicals 79614500ML
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Specifications
Phosphoric acid | |
85% | |
5 mmHg (25°C), 2.2 mmHg (20°C) | |
H3O4P | |
MFCD00011340 | |
1921286 | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
phosphoric acid | |
1004 | |
98g/mol |
7664-38-2 | |
1.685g/mL at 25°C | |
500 mL | |
H3PO4 | |
UN1805 | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
OP(=O)(O)O | |
97.994 | |
CHEBI:26078 | |
TraceSELECT™ |
Chemical Identifiers
7664-38-2 | |
97.994 | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
1004 | |
phosphoric acid |
H3O4P | |
MFCD00011340 | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
CHEBI:26078 | |
OP(=O)(O)O |
Safety and Handling
P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
RTECSNumber : TB6300000