Learn More
Phosphorus(III) chloride, 98% min, Thermo Scientific Chemicals
$65.74 - $283.05
Chemical Identifiers
CAS | 7719-12-2 |
---|---|
Molecular Formula | Cl3P |
Molecular Weight (g/mol) | 137.32 |
MDL Number | MFCD00011438 |
InChI Key | FAIAAWCVCHQXDN-UHFFFAOYSA-N |
Synonym | phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride |
PubChem CID | 24387 |
ChEBI | CHEBI:30334 |
IUPAC Name | trichlorophosphane |
SMILES | P(Cl)(Cl)Cl |
Description
Phosphorus(III) chloride is used as a reagent in organic synthesis. It is utilized in the conversion of primary alcohol and secondary alcohol to alkyl chlorides. It acts as a precursor to phosphorous pentachloride, phosphorous oxychloride, thiophosphoryl chloride and triphenylphosphine. It is also used to prepare phosphite esters, which is used Horner-Wadsworth-Emmons reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7719-12-2 | |
137.32 | |
FAIAAWCVCHQXDN-UHFFFAOYSA-N | |
24387 | |
trichlorophosphane |
Cl3P | |
MFCD00011438 | |
phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride | |
CHEBI:30334 | |
P(Cl)(Cl)Cl |
Specifications
76°C | |
7719-12-2 | |
Liquid | |
98% min. | |
MFCD00011438 | |
Miscible with benzene,carbon disulfide,ether,chloroform and carbon tetrachloride. | |
P(Cl)(Cl)Cl | |
137.32 | |
CHEBI:30334 | |
Acrid | |
95mm Hg at 20°C | |
Phosphorus(III) chloride |
°C to 111.8°C | |
Yellow | |
50 g | |
Cl3P | |
phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride | |
FAIAAWCVCHQXDN-UHFFFAOYSA-N | |
trichlorophosphane | |
24387 | |
137.33 | |
1.516 | |
1.574^2^1 |
Safety and Handling
Keep container tightly closed and dry. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H300+H330-H314-H373
DOTInformation : DOT Class: 6.1, Packaging Group: I
EINECSNumber : 231-749-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only