missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus pentoxide, 99+%, for analysis

CAS: 1314-56-3 | O5P2 | 141.94 g/mol

Supplier:  Thermo Scientific Chemicals 215755000

 Afficher plus de versions de ce produit

Numéro de catalogue. AC215755000


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
Spécifications

Spécifications

Phosphorus pentoxide
p.a.
White
500 g
O5P2
MFCD00011440
15, 7467
Solubility in water: 85% (20°C); reacts forming h3po4 with evol.of heat.
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
141.94
≥99%
Glass bottle
340.0°C to 360.0°C
1314-56-3
Very Deliquescing Powder
99+%
P2O5
01,871; 07,291; 10,319; 14,151; 15,267
Phosphoric anhydride
DLYUQMMRRRQYAE-UHFFFAOYSA-N
tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
141.94
Analytical
2.39

Identifiants chimiques

1314-56-3
141.94
DLYUQMMRRRQYAE-UHFFFAOYSA-N
tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
O5P2
MFCD00011440
Phosphoric anhydride
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 215-236-1

missing translation for 'rtecsNumber' : TH3945000

missing translation for 'tsca' : TSCA

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only