missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) oxybromide, 98% min

For Use As a Chemical Intermediate | CAS: 7789-59-5 | Br3OP | 286.685 g/mol

$86.67 - $774.97

Chemical Identifiers

CAS 7789-59-5
Molecular Formula Br3OP
Molecular Weight (g/mol) 286.685
MDL Number MFCD00036298
InChI Key UXCDUFKZSUBXGM-UHFFFAOYSA-N
Synonym phosphorus oxybromide, phosphoric tribromide, phosphoryl bromide, phosphoryl tribromide, phosphorusoxybromide, phosphorus v oxybromide, unii-h98h8y87qq, phosphorousoxybromide, phosphoroyl tribromide, pobr3
PubChem CID 24613
SMILES O=P(Br)(Br)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA5613506
View Documents
Thermo Scientific Chemicals
05613506
5 g
Each for $86.67
Only null left
 
AA5613514
View Documents
Thermo Scientific Chemicals
05613514
25 g
Each for $259.49
Only null left
 
AA5613522
View Documents
Thermo Scientific Chemicals
05613522
100 g
Each for $774.97
Only null left
 
Description

Description

As a chemical intermediate.Phosphorus(V) oxybromide is used as a brominating agent in the synthesis of 2,4,6-tribromopyrimidine and bromoheterocyclic compounds. It is used in the phosphorus flame retardant and phosphorus activators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-59-5
286.685
UXCDUFKZSUBXGM-UHFFFAOYSA-N
24613
Br3OP
MFCD00036298
phosphorus oxybromide, phosphoric tribromide, phosphoryl bromide, phosphoryl tribromide, phosphorusoxybromide, phosphorus v oxybromide, unii-h98h8y87qq, phosphorousoxybromide, phosphoroyl tribromide, pobr3
O=P(Br)(Br)Br

Specifications

189°C
7789-59-5
Lump
98% min.
MFCD00036298
Decomposes in water. Soluble in ether,carbon disulfide,benzene,chloroform and sulfuric acid.
O=P(Br)(Br)Br
24613
Moisture sensitive
Phosphorus(V) tribromide oxide
56°C
White to Yellow
5 g
Br3OP
phosphorus oxybromide, phosphoric tribromide, phosphoryl bromide, phosphoryl tribromide, phosphorusoxybromide, phosphorus v oxybromide, unii-h98h8y87qq, phosphorousoxybromide, phosphoroyl tribromide, pobr3
UXCDUFKZSUBXGM-UHFFFAOYSA-N
286.685
286.73
2.822
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : DOT Class: 8; Packaging Group: II

EINECSNumber : 232-177-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only