missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphorus(V) oxychloride, 99%, Thermo Scientific Chemicals

$99.15 - $241.05

Chemical Identifiers

CAS 10025-87-3
Molecular Formula Cl3OP
Molecular Weight (g/mol) 153.323
MDL Number MFCD00011443
InChI Key XHXFXVLFKHQFAL-UHFFFAOYSA-N
Synonym phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
PubChem CID 24813
ChEBI CHEBI:30336
SMILES O=P(Cl)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1052530
View Documents
Thermo Scientific Chemicals
1052530
250 g
Each for $99.15
 
AA10525A1
View Documents
Thermo Scientific Chemicals
010525A1
1 kg
Each for $241.05
 
Description

Description

Phosphorus(V) oxychloride is widely used as a dehydrating agent and a chlorinating agent. Useful for the conversion of amide to nitriles, Bischler-Napieralski reaction, Vilsmeier Haak reaction, and other reactions producing commercially important intermediates. Acts a source of liquid phosphorus in semiconductor industry. Manufacture of trialkyl phosphate esters and triarylphosphate esters, including commercially popular tricresyl phosphate,which is useful as a plasticizer, flame retardant, gasoline additive, hydrolytic fluid, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10025-87-3
153.323
XHXFXVLFKHQFAL-UHFFFAOYSA-N
24813
O=P(Cl)(Cl)Cl
Cl3OP
MFCD00011443
phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
CHEBI:30336

Specifications

105.8°C
10025-87-3
Liquid
99%
POCl3
UN1810
phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
XHXFXVLFKHQFAL-UHFFFAOYSA-N
153.323
CHEBI:30336
99%
1.46
1.645 g/mL
1.25°C
Colorless
250 g
Cl3OP
MFCD00011443
14,7349
Phosphorus(V) oxychloride is soluble in many organic solvents.
O=P(Cl)(Cl)Cl
24813
153.33
Pungent
Moisture sensitive
Phosphorus(V) oxychloride
Videos
Safety and Handling

Safety and Handling

Keep container tightly closed and dry. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P234-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H302-H314-H330-H335-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: PHOSPHORUS OXYCHLORIDE

EINECSNumber : 233-046-7

RTECSNumber : TH4897000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only