Motic™ Photo Eyepieces
Photo eyepiece to be used with the C-mount Photo Eyepiece Adapter.
$478.32 - $603.30
Specifications
For Use With (Application) | Attaching DSLR Camera To Microscope |
---|---|
For Use With (Equipment) | BA310MET Series, C-mount Photo Eyepiece Adapter |
Catalog Number | Mfr. No. | Magnification Power | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Magnification Power | Price | Quantity | |||||
01-674-242
|
Motic™
1101000300442 |
2.5X |
Each for $603.30
|
|
|||||
01-674-243
|
Motic™
1101000300452 |
4X |
Each for $478.32
|
|
|||||
Description
- Also requires C-mount Photo Eyepiece Adapter and T-Mount adapter from your camera's manufacturer
Specifications
Attaching DSLR Camera To Microscope | |
BA310MET Series, C-mount Photo Eyepiece Adapter |