Learn More
Phthaldialdehyde, 98%, Thermo Scientific Chemicals
$172.75 - $2308.29
Chemical Identifiers
CAS | 643-79-8 |
---|---|
Molecular Formula | C8H6O2 |
Molecular Weight (g/mol) | 134.13 |
MDL Number | MFCD00003335 |
InChI Key | ZWLUXSQADUDCSB-UHFFFAOYSA-N |
Synonym | o-phthalaldehyde, o-phthaldialdehyde, benzene-1,2-dicarboxaldehyde, 1,2-benzenedicarboxaldehyde, phthaldialdehyde, phthalic aldehyde, phthalic dialdehyde, phthalyldicarboxaldehyde, ortho-phthalaldehyde, o-phthaldehyde |
PubChem CID | 4807 |
ChEBI | CHEBI:70851 |
IUPAC Name | phthalaldehyde |
SMILES | O=CC1=CC=CC=C1C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1329909
|
Thermo Scientific Chemicals
A1329909 |
10 g |
Each for $172.75
|
|
||||||
AAA1329918
|
Thermo Scientific Chemicals
A1329918 |
50 g |
Each for $635.51
|
|
||||||
AAA1329930
|
Thermo Scientific Chemicals
A1329930 |
250 g |
Each for $2,308.29
|
|
||||||
Description
Phthaldialdehyde is used as a fluorescent reagent for checking the assay of amines present in proteins, peptides by capillary electrophoresis and chromatography. It is also used for cholesterol assay in the picomole range as well as for fluorometric determination of histamine, histidine and other amino acids. Poly(phthalaldehyde) is obtained by the polymerization reaction which is used to make a photoresist. The nitrogen by o-phthaldialdehyde assay (NOPA) is a method used in winemaking process to measure the completion of fermentation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhthaldialdehyde is used as a fluorescent reagent for checking the assay of amines present in proteins, peptides by capillary electrophoresis and chromatography. It is also used for cholesterol assay in the picomole range as well as for fluorometric determination of histamine, histidine and other amino acids. Poly(phthalaldehyde) is obtained by the polymerization reaction which is used to make a photoresist. The nitrogen by o-phthaldialdehyde assay (NOPA) is a method used in winemaking process to measure the completion of fermentation.
Solubility
Soluble in acetic acid and methanol. Slightly soluble in water.
Notes
Air, light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, amines and strong bases.
Chemical Identifiers
643-79-8 | |
134.13 | |
ZWLUXSQADUDCSB-UHFFFAOYSA-N | |
4807 | |
phthalaldehyde |
C8H6O2 | |
MFCD00003335 | |
o-phthalaldehyde, o-phthaldialdehyde, benzene-1,2-dicarboxaldehyde, 1,2-benzenedicarboxaldehyde, phthaldialdehyde, phthalic aldehyde, phthalic dialdehyde, phthalyldicarboxaldehyde, ortho-phthalaldehyde, o-phthaldehyde | |
CHEBI:70851 | |
O=CC1=CC=CC=C1C=O |
Specifications
643-79-8 | |
132°C (269°F) | |
C8H6O2 | |
10 g | |
878317 | |
o-phthalaldehyde, o-phthaldialdehyde, benzene-1,2-dicarboxaldehyde, 1,2-benzenedicarboxaldehyde, phthaldialdehyde, phthalic aldehyde, phthalic dialdehyde, phthalyldicarboxaldehyde, ortho-phthalaldehyde, o-phthaldehyde | |
ZWLUXSQADUDCSB-UHFFFAOYSA-N | |
phthalaldehyde | |
4807 | |
134.14 | |
o-Phthaldialdehyde |
53°C to 57°C | |
98% | |
MFCD00003335 | |
UN2923 | |
Air Sensitive | |
Soluble in acetic acid and methanol. Slightly soluble in water. | |
O=CC1=CC=CC=C1C=O | |
134.13 | |
CHEBI:70851 | |
98% | |
98% |
Safety and Handling
GHS H Statement
H300-H314-H318-H317
P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H301-H314-H317-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber : 211-402-2
RTECSNumber : TH6950000
TSCA : Yes
storageNote1 : Air sensitive
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only