Learn More
Phthalic anhydride, 99%, Thermo Scientific Chemicals
$39.62 - $39.62
Chemical Identifiers
CAS | 85-44-9 |
---|---|
Molecular Formula | C8H4O3 |
Molecular Weight (g/mol) | 148.12 |
MDL Number | MFCD00005918 |
InChI Key | LGRFSURHDFAFJT-UHFFFAOYSA-N |
Synonym | phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen |
PubChem CID | 6811 |
ChEBI | CHEBI:36605 |
SMILES | O=C1OC(=O)C2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1495522
|
Thermo Scientific Chemicals
A1495522 |
100 g |
Each for $39.62
|
|
|||||
Description
Phthalic anhydride is used in the preparation of primary amines. It is used as a precursor to prepare anthraquinone, phthalein, rhodamine, phthalocyanine, fluorescein and xanthene dyes. It serves as an important intermediate in the plastics industry, dyes, resins and pigments. It is used as a monomer for synthetic resins such as glyptal, alkyd resins and polyester resins. It plays a vital role in the large-scale production of plasticizers for plastics. Further, it acts as a curing agent for epoxy resins, anti-scaling agents, plating agents, surface treating agents, ion exchange agents, paint additives, adhesives, sealant chemicals and potential antibacterial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhthalic anhydride is used in the preparation of primary amines. It is used as a precursor to prepare anthraquinone, phthalein, rhodamine, phthalocyanine, fluorescein and xanthene dyes. It serves as an important intermediate in the plastics industry, dyes, resins and pigments. It is used as a monomer for synthetic resins such as glyptal, alkyd resins and polyester resins. It plays a vital role in the large-scale production of plasticizers for plastics. Further, it acts as a curing agent for epoxy resins, anti-scaling agents, plating agents, surface treating agents, ion exchange agents, paint additives, adhesives, sealant chemicals and potential antibacterial agent.
Solubility
Soluble in water, carbon disulfide, ethanol, acetone and benzene. Slightly soluble in ethyl ether.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases, strong reducing agents and strong acids.
Chemical Identifiers
85-44-9 | |
148.12 | |
LGRFSURHDFAFJT-UHFFFAOYSA-N | |
6811 | |
O=C1OC(=O)C2=CC=CC=C12 |
C8H4O3 | |
MFCD00005918 | |
phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen | |
CHEBI:36605 |
Specifications
85-44-9 | |
1.53 | |
152°C (305°F) | |
C8H4O3 | |
100 g | |
118515 | |
14,7372 | |
Soluble in water,carbon disulfide,ethanol,acetone and benzene. Slightly soluble in ethyl ether. | |
O=C1OC(=O)C2=CC=CC=C12 | |
148.12 | |
CHEBI:36605 | |
99% |
130°C to 134°C | |
295°C (sublimation) | |
Weak, Characteristic | |
MFCD00005918 | |
UN2214 | |
Moisture sensitive | |
phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen | |
LGRFSURHDFAFJT-UHFFFAOYSA-N | |
1,3-dihydro-2-benzofuran-1,3-dione | |
6811 | |
148.12 | |
Phthalic anhydride |
Safety and Handling
GHS H Statement
H334-H318-H302-H335-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P342+P311-P362-P501c
H302-H315-H317-H318-H334-H335-H500
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: PHTHALIC ANHYDRIDE
EINECSNumber : 201-607-5
RTECSNumber : TI3150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only