missing translation for 'onlineSavingsMsg'
Learn More

Picolinic acid N-oxide, 97%, Thermo Scientific™

Chemical Identifiers

CAS 824-40-8
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006198
InChI Key FHYMLBVGNFVFBT-UHFFFAOYSA-N
Synonym picolinic acid n-oxide, 2-carboxypyridin-1-ium-1-olate, pyridine-2-carboxylic acid 1-oxide, picolinic acid 1-oxide, 2-carboxypyridine n-oxide, 2-pyridinecarboxylic acid, 1-oxide, chembl73173, 2-pyridinecarboxylicacid, 1-oxide, picolinate n-oxide, 1-oxopicolinic acid
PubChem CID 69986
IUPAC Name 1-oxidopyridin-1-ium-2-carboxylic acid
SMILES C1=CC=[N+](C(=C1)C(=O)O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC131180050
SDS View Documents
Thermo Scientific Chemicals
131180050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC131180250
SDS View Documents
Thermo Scientific Chemicals
131180250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

824-40-8
139.11
FHYMLBVGNFVFBT-UHFFFAOYSA-N
69986
C1=CC=[N+](C(=C1)C(=O)O)[O-]
C6H5NO3
MFCD00006198
picolinic acid n-oxide, 2-carboxypyridin-1-ium-1-olate, pyridine-2-carboxylic acid 1-oxide, picolinic acid 1-oxide, 2-carboxypyridine n-oxide, 2-pyridinecarboxylic acid, 1-oxide, chembl73173, 2-pyridinecarboxylicacid, 1-oxide, picolinate n-oxide, 1-oxopicolinic acid
1-oxidopyridin-1-ium-2-carboxylic acid

Specifications

824-40-8
96% min. (HPLC)
C6H5NO3
5g
FHYMLBVGNFVFBT-UHFFFAOYSA-N
1-oxidopyridin-1-ium-2-carboxylic acid
69986
97%
Authentic
Glass bottle
MFCD00006198
picolinic acid n-oxide, 2-carboxypyridin-1-ium-1-olate, pyridine-2-carboxylic acid 1-oxide, picolinic acid 1-oxide, 2-carboxypyridine n-oxide, 2-pyridinecarboxylic acid, 1-oxide, chembl73173, 2-pyridinecarboxylicacid, 1-oxide, picolinate n-oxide, 1-oxopicolinic acid
C1=CC=[N+](C(=C1)C(=O)O)[O-]
139.11
139.11
Picolinic acid N-oxide, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

EINECSNumber : 212-528-0

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only