Thermco™ PicoLite Disposable USB Temperature Cold Chain Portable Data-Logger
Used for monitoring and recording temperatures and humidity over long periods of time. Thermco™ Products PicoLite Disposable USB Temperature Cold Chain Portable Data-Logger is low cost, compact, and easy-to-use.
Supplier:  Thermco™ ACCDPICOLITE
Catalog No. 04500002
Explore available promotions
Includes: DataSuite software
Description
Description
  • Features built-in temperature sensor, activation button
  • Requires control over quality of perishable goods
  • Offers plug and record solution for every application
  • Low cost, one-trip, disposable logger for unbroken cold chain
Certifications
Can be N.I.S.T. Certified
Warranty
1 Year
Specifications
Specifications
Datalogger, Disposable USB Temperature Cold Chain Portable
CR2032
CE, EN12830, IP67 compliant
0.5°C
74 x 30 x 13 mm
LED, Bi-color for sampling and alarm indication
Up to 6 months depending on operating conditions
USB 2.0
-40°C to +40°C
15 g