Picric Acid, Crystal, Reagent, ACS, Spectrum™ Chemical
P1145, 88-89-1, C6H2(NO2)3OH
Supplier:  Spectrum Chemical Mfg Cor P114525GM
 View more versions of this product
Catalog No. 18605212
Explore available promotions
Description
Description
Spectrum™ Chemical Picric Acid, Crystal, Reagent, ACS , also known as 2,4,6-trinitrophenol or TNP. It is a yellow crystalline solid and one of the most acidic phenols. As a highly nitrated compound, picric acid is highly explosive (like TNT). Apart from it's use as an explosive, it also has some uses in organic chemistry for the preparation of crystalline salts of organic bases. It is also used in clinical chemistry for the Jaffe reaction (a colorimetric method to determine creatinine levels in blood and urine).
Specifications
Specifications
88-89-1
C6H3N3O7
OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O
229.10
25 g
1
OXNIZHLAWKMVMX-UHFFFAOYSA-N
2,4,6-trinitrophenol
ACS
Amber Glass Bottle
Chemical Identifiers
88-89-1
229.10
2,4,6-trinitrophenol
C6H3N3O7
OXNIZHLAWKMVMX-UHFFFAOYSA-N
OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O