missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Picrotoxin, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 131210250

Catalog No. AC131210250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Picrotoxin
Authentic
Glass Bottle
25g
− 32.00
Solubility in water: 1g/5mL (100°C). Other solubilities: soluble in boiling alcohol (1gr/3mL),dilute acids,and alkalies; sparingly soluble in ether and ccl4,readily soluble in strong ammonia water and in aq.,solns of naoh.,1gr/13.5 mL in 95% ethan
CC(=C)C1C2C3C4(C(C1C(=O)O2)(CC5C4(O5)C(=O)O3)O)C.CC12C3C4C(C(C1(CC5C2(O5)C(=O)O3)O)C(=O)O4)C(C)(C)O
131846146
98%
124-87-8
97.5% min. (Mixture of Picrotoxinin and Picrotin) (HPLC)
C30H34O13
15,7527
picrotoxin
VJKUPQSHOVKBCO-DMRKGALJSA-N
602.58
602.58
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 204-716-6

Recommended Storage : 4°C

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only