Thermo Scientific™ Plate Seal for Pierce™ 96-well Microdialysis Plates
Simultaneously process 1 to 96 samples with 10 to 100 μL volumes with this convenient and disposable rapid dialysis system that is automation compatible.
Supplier:  Thermo Scientific™ 88269
Catalog No. PI88269
Explore available promotions
Description
Description
The Thermo Scientific™ Pierce 96-well Microdialysis Plate, 10K MWCO is a convenient and disposable rapid dialysis system for the simultaneous processing of 1 to 96 samples with volumes 10 to 100 μL and is compatible with automation systems.
Thermo Scientific Pierce Plate Seal for Pierce 96-well Microdialysis Plates is an adhesive coated plastic sheet, perforated for use with an 8-well strip microdialysis plate. Sufficient For: 12 x 8 microdialysis plate wells.
These plate seals are made of high-quality polypropylene and help protect samples from contamination and evaporation, especially during long-term storage.
Specifications
Specifications
12 x 8 microdialysis plate wells
Microdialysis Plates
1 Sheet
Upon receipt store at room temperature. Product shipped at ambient temperature.
Slide-A-Lyzer™