missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Biotin Quantitation Kit

HABA dye reagent and biotinylated protein standard for convenient colorimetric determination of biotinylation levels in antibodies and other proteins.

Supplier:  Thermo Scientific™ 28005

Catalog No. PI28005

Explore available promotions

Description

Description

Thermo Scientific™ Pierce Biotin Quantitation Kit includes aliquots of HABA reagent and biotinylated protein standard for convenient colorimetric determination of biotinylation levels in labeled antibodies and other proteins.

Determine the molar ratio of biotin incorporated into a protein using the HABA-avidin method. The HABA dye (4'-hydroxyazobenzene-2-carboxylic acid) binds to avidin to produce a yellow-orange colored complex that absorbs at 500nm. Free biotin present in solution with this avidin-HABA complex will displace the HABA dye and cause the absorbance to decrease.The amount of biotin present can be calculated directly from the decreased absorbance at 500nm. The Thermo Scientific EZ Biotin Quantitation Kit contains pre-measured doses of the Avidin-HABA mixture and biotinylated horseradish peroxidase positive control to simplify reagent preparation and minimize the amount of waste generated. The convenient assay can be performed either in a cuvette or in a microplate.

Highlights:

  • HABA-avidin complex can be used over a wide range of pH and salt concentrations
  • Amount of biotin can be calculated directly from the decreased absorbance at 500nm complexing with the HABA dye

Eliminate the need for cumbersome assay equations with a convenient HABA calculator found at www.piercenet.com/HABA/habacalc.cfm

Includes:

HABA/Avidin Premix, 24 tubes; Biotinylated HRP, 12 tubes

Specifications

Specifications

Biotin Quantitation Kit
Sufficient For: Testing biotinylation of 12 protein samples
• HABA-Avidin Premix, 24 tubes
• Biotinylated Horseradish Peroxidase, 5 mg

Store at 4°C.
Biotin
12 Tests
Pierce™
Antibodies, Proteins
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.