Learn More
Thermo Scientific™ Compat-Able™ BCA Protein Assay Kit
Precipitation reagents to eliminate interfering substances from protein assay samples to make them compatible with BCA and Coomassie-Bradford assays.
Supplier: Thermo Scientific™ LSG23229
Description
Thermo Scientific™ Compat-Able Protein Assay Preparation Set eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.
The Compat-Able Reagents remove salts, detergents, reducing agents and other substances from protein samples to eliminate interference with protein assays. The protocol selectively precipitates all of the protein, thereby allowing the non-protein components to be easily decanted and removed. The purified, precipitated protein is redissolved in water, making it ready for accurate determination of the protein concentration by standard protein assay methods. The procedure is accomplished in less than 10 minutes with just a few simple steps. The Compat-Able Preparation Set is available individually or conveniently bundled with either the Thermo Scientific Pierce BCA Protein Assay Kit or the Thermo Scientific Pierce Coomassie Plus Protein Assay Kit.Highlights:
Eliminate incompatibilities – removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer
Simple and fast – prepare samples in less than 10 minutes with four simple steps
Convenient – reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results
Scalable – microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs
Validated – tested and offered together with BCA and Coomassie Plus Protein Assay Kits; protocol integrates control and normalization procedures
Includes:
Set contains two bottled reagents (Reagent 1 and Reagent 2); Complete kits contain the Reagent Set plus components of the respective protein assay
Spécifications
Sufficient For: Treating up to 500 protein assay samples and performing 250 tube assays or 2500 microplate assays • Compat-Able Reagent 1, 250 mL • Compat-Able Reagent 2, 250 mL • BCA Reagent A, 500 mL • BCA Reagent B, 25 mL • Albumin Standard Ampules, 2 mg/mL, 10 x 1 mL |
|
BCA Assay | |
Compat-Able™ | |
Not Target-Specific | |
Absorbance, Solution-based Detection, Incompatibility Elimination | |
1 L |
Compat-Able BCA Protein Assay Kit | |
Colorimetric | |
Protein Quantitation Assay | |
250 Tube Assays or 2500 Microplate Assays | |
Spectrophotometer, Microplate Reader |
For Research Use Only. Not for use in diagnostic procedures.