missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Bradford Plus Protein Assay Reagent

Brown-to-purple (A595nm), ready-to-use, fast reducing agent-compatible assay reagent to measure total protein concentration vs. protein standard.

Supplier:  Thermo Scientific™ LSG23238

Catalog No. PI23238


Explore available promotions

Description

Description

Thermo Scientific™ Pierce Coomassie Plus Protein Assay is a high-performance Bradford assay reagent — one of the fastest, simplest and most popular protein quantitation methods available.

The Pierce Coomassie Plus Assay Reagent is a single, ready-to-use solution for measuring protein concentration. Simply add the reagent to equal volumes of samples and standards, mix, then measure the absorbance at 595nm. The assay costs only pennies per sample and can be performed in either test tube or microplate format. The Pierce Coomassie Plus Assay Reagent provides increased linearity of response and only half the protein-to-protein variability of other commercial Bradford assay formulations.

Highlights:

  • Colorimetric – measure with a standard spectrophotometer or plate reader (595nm)

  • Easy to use – single reagent; no working reagent preparation required

  • Fast – almost immediate color development; add, mix, and read results

  • Broad range – detects protein concentration in the range 1 to 1500μg/mL

  • Better – improved linearity and response-uniformity compared to traditional Bradford formulations

  • Flexible – microplate and cuvet protocols provided with the instructions and adaptable to several target working ranges

 

Specifications

Specifications

Store at 4°C.
Bradford Assay
Pierce™
Not Target-Specific
Spectrophotometer, Microplate Reader
Pierce Bradford Plus Protein Assay Reagent
Colorimetric
Protein Quantitation Assay
Solution-based Detection, Absorbance
300 mL
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.