Thermo Scientific™ Pierce™ DSG, No-Weigh™ Format
$248.36 - $258.15
Specifications
Chemical Reactivity Amine-Amine
Cleavable No
Molecular Weight (g/mol) 326.26
PEGylated No
Spacer Arm Length 7.7 Å
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PIA35392
View Documents
Thermo Scientific™
A35392
10 x 1 mg N/A
PI20593
View Documents
Thermo Scientific™
20593
50 mg N/A
Description
Description
Thermo Scientific™ Pierce DSG is disuccinimidyl glutarate, a simple homobifunctional crosslinker based on the amine-reactive N-hydroxysuccinimide (NHS) ester group.
Highlights:
  • Can increase crosslinking efficiency compared to DSS in some applications
  • Reactive groups: NHS ester (homobifunctional)
  • Reactive toward: amino groups
Specifications
Specifications
Amine-Amine
326.26
7.7 Å
Chemical Labeling
NHS Ester
Powder
Standard, Single-use
Crosslinker
No
No
Yes
Homobifunctional
Short (<10 Å)
DMF, DMSO
No
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
For Research Use Only. Not for use in diagnostic procedures.