missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ DTME (dithiobismaleimidoethane)

Crosslink sulfhydryls to sulfhydryls (cysteine residues) with this long, cleavable crosslinker composed of maleimide groups and 11-atom (13.3A) disulfide spacer arm.

Supplier:  Thermo Scientific™ LSG22335

Catalog No. PI22335

Explore available promotions

Description

Description

Thermo Scientific™ Pierce DTME is a mid-length, maleimide crosslinker for reversible covalent conjugation between sulfhydryl groups (e.g., protein or peptide cysteines) by reduction of the disulfide bond in the center of the spacer arm.

Highlights:

  • Cleavable by reducing agents
  • Reactive groups: maleimide (homobifunctional)
  • Reactive toward: sulfhydryl groups
Specifications

Specifications

Sulfhydryl-Sulfhydryl
DTME
No
Upon receipt store desiccated at 4°C.
Ambient
Chemical Labeling
Maleimide
Solid
DMF, DMSO
No
By Thiols
312.36
13.3 Å
Yes
Pierce™
Homobifunctional
Medium (10 to 30 Å)
50 mg
Standard
Crosslinker
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.