missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ DTT (Dithiothreitol)
Reduce disulfides in biological buffers and sample loading buffers with pure, crystalline dithiothreitol (DTT), the most popular protein disulfide reducing agent.
Supplier: Thermo Scientific™ 20290
Description
Dithiothreitol is a popular reducing agent for many protein research applications, including reducing polyacrylamide electrophoresis (SDS-PAGE).
Dithiothreitol is a popular reducing agent for many protein research applications, including reducing polyacrylamide electrophoresis (SDS-PAGE). 5g glass bottle.Specifications
Pierce DTT, Cleland's Reagent | |
Pierce™ | |
5 g |
Store at 4°C. | |
Powder | |
Dithiothreitol |
For Research Use Only. Not for use in diagnostic procedures.