missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ ELISA Wash Buffer (30X)
Concentrated wash buffer for efficient use of storage in the lab
Supplier: Thermo Scientific™ N503
Description
- Type: Reagent
- Format: 30X Concentrate
Specifications
Wash Buffer | |
Liquid | |
Store at -20°C. |
50 mL | |
ELISA |
For Research Use Only. Not for use in diagnostic procedures.