missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Ethidium Bromide Solution (0.625 mg/mL)

Visualize DNA fluorescently in a variety of molecular biology applications using these dilute (0.625mg/mL) or concentrated (10mg/mL) ethidium bromide solutions.

Supplier:  Thermo Scientific™ LSG17896

Catalog No. PI17896


Explore available promotions

Description

Description

Thermo Scientific™ Pierce Ethidium Bromide is available as both dilute (0.625mg/mL) and more concentrated (10mg/mL) solutions to accommodate most molecular biology applications for fluorescent DNA visualization.

Thermo Scientific™ Pierce Ethidium Bromide is available as both dilute (0.625mg/mL) and more concentrated (10mg/mL) solutions to accommodate most molecular biology applications for fluorescent DNA visualization.

Highlights:

0.625mg/mL EtBr Solution:

  • Each drop contains 25μg of ethidium bromide and is ideal for a 50mL agarose gel
  • After adding one drop of solution per 50mL of gel, the final concentration of EtBr will be 0.5μg/mL, which is the recommended concentration for electrophoresis of nucleic acids
  • Can also be used as the running buffer or for staining gels after electrophoresis

10mg/mL EtBr Solution:

  • A safe and convenient way to use ethidium bromide while minimizing exposure
Specifications

Specifications

Store at room temperature, protected from light.
Fluorescence
Ethidium Bromide Solution
RNA, DNA
In-Gel Detection
Ethidium bromide
5 mL
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.