missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Formic Acid, LC-MS Grade

A high-purity solvent supplied in glass ampules as a convenient, contamination-free alternative for preparing elution solvents for HPLC separations of protein and peptides.

Supplier:  Thermo Scientific™ 28905

Catalog No. PI28905


Explore available promotions

Description

Description

Thermo Scientific™ Formic Acid is high-purity solvent supplied in glass ampules as a convenient, contamination-free alternative for preparing elution solvents for HPLC separations of protein and peptides.

Formic acid is a component found in reverse-phase mobile phases to provide protons for LC/MS analysis. The presence of a low concentration of formic acid in the mobile phase is also known to improve the peak shapes of the resulting separation. Unlike trifluoroacetic acid (TFA), formic acid is not an ion-pairing reagent, and it does not suppress MS ionization of polypeptides when used as a mobile phase component.

Ordering Alerts:

  • X in the ordering table indicates that hazardous shipping charges apply.

Storage Requirements:

  • Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Specifications

Specifications

Formic acid
Formic Acid
Liquid
10 x 1 mL
Pierce™
Liquid
98 to 100%
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area
Solvent (Pure)
99% Formic Acid
LC/MS
Videos
SDS
Documents

Documents

Promotions

Promotions