Learn More
Thermo Scientific™ Formic Acid, LC-MS Grade
A high-purity solvent supplied in glass ampules as a convenient, contamination-free alternative for preparing elution solvents for HPLC separations of protein and peptides.
$356.39 - $356.39
Specifications
Item Name | Formic Acid |
---|---|
Form | Liquid |
Reagent Type | Solvent (Pure) |
Product Line | Pierce™ |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PI85178
|
Thermo Scientific™
T85178AD |
50 mL |
Each for $356.39
|
|
|||||
Description
Thermo Scientific™ Formic Acid is high-purity solvent supplied in glass ampules as a convenient, contamination-free alternative for preparing elution solvents for HPLC separations of protein and peptides.
Formic acid is a component found in reverse-phase mobile phases to provide protons for LC/MS analysis. The presence of a low concentration of formic acid in the mobile phase is also known to improve the peak shapes of the resulting separation. Unlike trifluoroacetic acid (TFA), formic acid is not an ion-pairing reagent, and it does not suppress MS ionization of polypeptides when used as a mobile phase component.
Highlights:
- Prescored, nitrogen-flushed, amber glass to protect formic acid from light and moisture
- 99% purity for consistent LC baselines and no interference introduced into LC and mass spectrometry applications
- Convenient format simplifies preparation of gradient and isocratic mobile phases containing 0.1% (v/v) formic acid in water or acetonitrile
- Contents of a single vial in a final volume of 1L solvent yields a mobile phase of the most common formic acid concentration
Ordering Alerts:
- X in the ordering table indicates that hazardous shipping charges apply.
Storage Requirements:
- Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Specifications
Formic Acid | |
Liquid | |
Solvent (Pure) | |
Pierce™ |
For Research Use Only. Not for use in diagnostic procedures.