Learn More
Thermo Scientific™ Guardian™ Peroxidase Conjugate Stabilizer/Diluent
Preserve diluted (1-1,000ng/mL) HRP conjugates for more than six months at room temperature without significant loss of activity using Guardian Peroxidase Conjugate Stabilizer/Diluent.
$325.39 - $846.87
Specifications
Product Type | Peroxidase Conjugate Stabilizer/Diluent |
---|---|
Product Line | Guardian™ |
Reagent Type | Diluent, Stabilizing Reagent |
Content And Storage | Upon receipt store at room temperature or 4°C. |
Form | Liquid |
Description
Thermo Scientific™ Guardian Peroxidase Conjugate Stabilizer/Diluent preserves diluted (1-1,000ng/mL) HRP conjugates for more than six months at room temperature without significant loss of activity.
Assay-compatible - simply add your favorite blocking buffer to create the ideal diluent for your HRP-based ELISA system
Convenient - store ready-to-use dilutions (1:1,000-1:100,000) that maintain enzyme activity in the refrigerator without aliquoting
Specifications
Peroxidase Conjugate Stabilizer/Diluent | |
Diluent, Stabilizing Reagent | |
Liquid |
Guardian™ | |
Upon receipt store at room temperature or 4°C. |
For Research Use Only. Not for use in diagnostic procedures.