missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ Heptafluorobutyric Acid (HFBA), HPLC Grade
Bottles and ampules of 99.5%-pure HFBA (MW 214.04, CAS 375-22-4), valid for use as an ion-pairing agent in reverse-phase peptide separations (HPLC).
Supplier: Thermo Scientific™ 53104
Description
Thermo Scientific™ Pierce Heptafluorobutyric Acid (HFBA) is manufactured and tested to meet strict specifications that ensure superior performance for use as an ion-pairing agent in reverse-phase peptide separations.
Highlights:
- Typical purity is 99.7% by GC; <0.1% water
- Sequencing reagent for classical and automated Edman degradation of peptides and proteins
- Density: 1.645
- B.P. 120°C
- Packaged under nitrogen in amber glass ampules or bottles
- Clear, colorless liquid
Ordering Information:
X in the ordering table indicates that hazardous shipping charges apply.
Specifications
Heptafluorobutyric Acid (HFBA) | |
Liquid | |
10 x 1 mL | |
Pierce™ |
Store at room temperature. | |
Ion Pairing Reagent | |
99.5% HFBA |
For Research Use Only. Not for use in diagnostic procedures.