Learn More
Thermo Scientific™ Pierce™ High Capacity Endotoxin Removal Spin Columns, 1 mL
Remove endotoxins (lipopolysaccharides or LPS) from samples before mass spectrometry using spin columns of poly(e-lysine) ligand on beaded cellulose.
Supplier: Thermo Scientific™ 88277
Description
Reduce endotoxin levels in protein samples by greater than 99% in one hour while maximizing protein recovery with Thermo Scientific™ Pierce High Capacity Endotoxin Removal Resin.
The Pierce High Capacity Endotoxin Removal Resin combines porous cellulose beads and an FDA-approved food preservative, poly(epsilon-lysine), as an affinity ligand to selectively bind endotoxins. Endotoxin levels in protein samples are reduced by greater than 99% in as little as 1 hour using our spin cup format, and protein recovery is ≥85%. The modified polylysine affinity ligand eliminates the toxicity concerns associated with alternative technologies using polymixin B ligands and sodium deoxycholate buffers. Pierce High capacity Endotoxin Removal Resin is available as a slurry to pack a custom column that can be re-used at least 5 times with no loss in endotoxin removal efficiency. This resin is also available in a new single-use spin column format optimized for different sample volumes.Highlights:
High capacity bind up to 2,000,000 EU/mL to eliminate at least 99% of endotoxins
Durable re-use resin up to 10 times with no loss in performance
Selective recover at least 85% of your protein sample
High performance compatible with FDA guidelines by reducing final EU concentration to less than 5 EU/mL
Fast our new spin cup format enables endotoxin depletion within 1 hour
Cleansingle-use spin columns avoid cross contamination of samples
Optimized spin cups are optimized for different sample volumes
Economical large-volume discounts available
Requires:
Endotoxin-free water
Recommended for:
Eliminate endotoxins from protein or antibody samples prior to cell transfection or animal injection
Specifications
Up to 2,000,000 EU/mL | |
Upon receipt store at 4°C. | |
Pierce™ | |
Endotoxin Removal | |
Proprietary Ligand | |
Affinity, Cellulose Resin |
22mL-capacity centrifuge columns containing 1mL resin | |
Liquid | |
25 Columns | |
Spin Column | |
Spin Column |
For Research Use Only. Not for use in diagnostic procedures.