missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ 16% Formaldehyde (w/v), Methanol-free

Vials of high-quality, methanol-free formaldehyde, a general IHC fixative and reversible amine-crosslinking agent for protein and nucleic acid samples.

$130.29 - $186.16

Specifications

Type Formaldehyde
Product Line Pierce
Format Liquid
Reagent Type Fixation Reagent
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PI28906
View Documents
Thermo Scientific™
28906
10 x 1 mL
Each for $130.29
 
PI28908
View Documents
Thermo Scientific™
LSG28908
10 x 10 mL
Each for $186.16
 
Description

Description

Thermo Scientific™ Pierce Formaldehyde Ampules are vials of high-quality, methanol-free formaldehyde for use as a general immunohistochemistry fixative and reversible amine-crosslinking agent for protein and nucleic acid tissue samples.

Formaldehyde is a highly reactive, cell-permeable agent that is used as a reversible cross-linking agent for proteins and nucleic acids within the cell or as a general cell-fixing agent for imaging-based applications. Cell permeability allows the reagent to readily enter living cells, enabling intracellular applications, such as protein interaction discovery or bioimaging. Formaldehyde produces reversible cross-links with protein and nucleic acids by coupling neighboring primary amines. Cross-links can be reversed simply by heating the sample.

Highlights:

  • Pure and stable - prepared directly from pure granular paraformaldehyde and sealed in ampules under an inert atmosphere, ensuring that it is methanol-free and resistant to oxidation

  • Versatile - use to cross-link molecules in protein-protein and protein-nucleic acid interaction discovery, bioimaging and flow cytometry

  • Aliquot packaging - choose 1mL or 10mL ampules suitable for low- and high-volume applications ranging from culture-targeted cell fixing to in-cell protein cross-linking

  • Convenient starting concentration - 16% (w/v) formaldehyde solution in ultrapure water simplifies dilution to the required working concentration

Specifications

Specifications

Formaldehyde
Liquid
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Pierce
Fixation Reagent
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.