Thermo Scientific™ Micro BCA™ Reagent C (MC)
Special 3-component version of our popular BCA Reagents to measure total protein concentration of dilute protein solutions (0.5 to 20 micrograms/mL).
Supplier:  Thermo Scientific™ LSG23234
Catalog No. PI23234
Explore available promotions
Description
Description
Thermo Scientific™ Pierce Micro BCA Protein Assay uses a concentrated form of the BCA Reagents to accurately measure dilute protein solutions (0.5 to 20μg/mL).
Additional Information and Online Ordering
The Micro BCA Protein Assay Kit is a specialized version of the popular Pierce BCA Protein Assay for determining the protein concentration of dilute samples. Mixing together the three Micro BCA Reagents results in a working solution that is sufficiently concentrated to measure protein when mixed with an equal volume of sample. The result is an assay for accurately measuring 0.5 to 20μg/mL protein solutions. The assay is exceptionally linear and exhibits very low levels of protein-to-protein variability.
Highlights:
  • Sensitive – accurately detect down to 0.5μg/mL (2μg/mL in microplate format)
  • High linearity – linear working range for BSA equals 0.5 to 20μg/mL
  • Colorimetric – measure with a standard spectrophotometer or plate reader (562nm)
  • Compatible – unaffected by typical concentrations of most ionic and nonionic detergents
  • Convenient – microplate and cuvette protocols provided with the instructions
  • Stable – kit stable at room temperature; prepared working reagent stable for to 24 hours
Includes:
Three Micro BCA Reagents (MA, MB, MC) and 10 ampules of Albumin Standard (2mg/mL)
Specifications
Specifications
Upon receipt store product at room temperature. Product shipped at ambient temperature.
BCA Assay
Micro BCA™, Pierce™
Not Target-Specific
Solution-based Detection, Absorbance
12 mL
Pierce Micro BCA Reagent C (MC)
Colorimetric
Protein Quantitation Assay
3200 Microplate Assays
Spectrophotometer, Microplate Reader
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.