missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Micro BCA™ Protein Assay Kit

Special 3-component version of our popular BCA Reagents to measure total protein concentration of dilute protein solutions (0.5 to 20 micrograms/mL).

Supplier:  Thermo Scientific™ LSG23235

Catalog No. PI23235


Explore available promotions

Description

Description

Thermo Scientific™ Pierce Micro BCA Protein Assay uses a concentrated form of the BCA Reagents to accurately measure dilute protein solutions (0.5 to 20μg/mL).

The Micro BCA Protein Assay Kit is a specialized version of the popular Pierce BCA Protein Assay for determining the protein concentration of dilute samples. Mixing together the three Micro BCA Reagents results in a working solution that is sufficiently concentrated to measure protein when mixed with an equal volume of sample. The result is an assay for accurately measuring 0.5 to 20μg/mL protein solutions. The assay is exceptionally linear and exhibits very low levels of protein-to-protein variability.

Highlights:

  • Sensitive – accurately detect down to 0.5μg/mL (2μg/mL in microplate format)

  • High linearity – linear working range for BSA equals 0.5 to 20μg/mL

  • Colorimetric – measure with a standard spectrophotometer or plate reader (562nm)

  • Compatible – unaffected by typical concentrations of most ionic and nonionic detergents

  • Convenient – microplate and cuvette protocols provided with the instructions

  • Stable – kit stable at room temperature; prepared working reagent stable for to 24 hours

Includes:

Three Micro BCA Reagents (MA, MB, MC) and 10 ampules of Albumin Standard (2mg/mL)

Specifications

Specifications

Sufficient For: 480 tube assays or 3200 microplate assays
• Micro BCA Reagent A (MA), 240 mL
• Micro BCA Reagent B (MB), 240 mL
• Micro BCA Reagent C (MC), 12 mL
• Albumin Standard Ampules, 2 mg/mL, 10 x 1 mL

Store at room temperature.
BCA Assay
Micro BCA™, Pierce™
Not Target-Specific
Solution-based Detection, Absorbance
500 mL
Pierce Micro BCA Protein Assay Kit
Colorimetric
Protein Quantitation Assay
480 Tube Assays or 3200 Microplate Assays
Spectrophotometer, Microplate Reader
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.