Learn More
Thermo Scientific™ PageBlue™ Protein Staining Solution
A sensitive formulation of colloidal coomassie G-250 dye for endpoint staining of proteins in polyacrylamide gels and on PVDF membranes without methanol or acetic acid.
Supplier: Thermo Scientific™ 24620
Description
Thermo Scientific Pierce PageBlue Protein Staining Solution is a sensitive, economical formulation of colloidal coomassie G-250 dye for endpoint staining of proteins in polyacrylamide gels and on PVDF membranes without methanol or acetic acid.
The simple PageBlue Protein Staining Solution protocol is sensitive, efficient and eliminates the concern of over staining gels even after overnight staining. This protein stain delivers a dynamic range of 5 to 500ng, which is approximately 10 times more sensitive than traditional coomassie R-250-based dyes. Furthermore, PageBlue Protein Staining Solution is economical because it can be reused up to three times without any loss in sensitivity.Highlights:
Easy-to-use – simply soak the gel in the ready-to-use solution and observe stained bands without destaining
Fast – 25 to 40 minute protocol
Safe – does not contain methanol or acetic acid
Dynamic range – linear dynamic range of 5 to 500ng
Economical – can be reused up to 3 times
Recommended for:
Visualize and quantify proteins separated in 1D, 2D and IEF polyacrylamide gels; Stain proteins after transfer onto PVDF membranes; Stain gels prior to subsequent silver staining or mass spectrometry analysis
Specifications
Upon receipt store product at room temperature. Product shipped at ambient temperature | |
Colorimetric | |
Protein Staining Solution | |
1 L |
In-Blot Detection, In-Gel Detection | |
Coomassie | |
PageBlue™ | |
Protein |
For Research Use Only. Not for use in diagnostic procedures.