Thermo Scientific™ Pierce™ Premium Grade DSP (dithiobis(succinimidyl propionate))
Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this popular amine-reactive, cleavable crosslinker.
Supplier:  Thermo Scientific™ PG82081
 View more versions of this product
Catalog No. PIPG82081
Explore available promotions
Description
Description
Optimize applications where product integrity and risk minimization are paramount with the specially characterized Thermo Scientific™ Pierce™ Premium Grade DSP, our highest quality formulation of this popular amine-reactive, cleavable crosslinker.
Thermo Scientific Pierce Premium Grade DSP is our highest quality formulation of this popular amine-reactive, cleavable crosslinker, specially characterized for applications where product integrity and risk minimization are paramount.
Compared to the standard grade product, Premium Grade DSP provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. DSP is dithiobis(succinimidyl propionate), an amine-reactive crosslinker having NHS-ester reactive ends and a cleavable disulfide bond in its spacer arm. DSP contains an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of an 8-carbon spacer arm. NHS esters react with primary amines at pH 7-9 to form stable amide bonds, along with release of the N-hydroxy-succinimide leaving group. Proteins, including antibodies, generally have several primary amines in the side chain of lysine (K) residues and the N-terminus of each polypeptide that are available as targets for NHS-ester crosslinking reagents.
Highlights:
  • DSP – a thiol-cleavable, homobifunctional and amine-reactive crosslinking agent
  • High quality – identity and purity confirmed by several tests, including quantitative NMR
  • Product integrity – enhanced level of testing and characterization compared to standard grade
  • Lot retention – ample supply of past lots retained to ensure future process testing
  • Change management – Change Control Notification (CCN) service
  • Consistent manufacture – batch-specific manufacturing documentation review
Applications:
  • Chemical crosslinking of intracellular proteins prior to cell lysis and immunoprecipitation
  • “Fix” protein interactions to allow identification of weak or transient protein interactions
  • Fix tissues for immunostaining without damaging RNA
  • Protein crosslinking to create bioconjugates via single-step reactions
  • Immobilize proteins onto amine-coated surfaces
Specifications
Specifications
Amine-Amine
Dithiobis(succinimidylpropionate); powder solid
No
12.0 Å
Yes
Pierce™
Homobifunctional
Medium (10 to 30 Å)
1 g
Standard, Single-use, Premium-grade
Crosslinker
By Thiols
404.42
2.47L of typical 1X (1mM) reaction solution
Upon receipt store desiccated at 4°C.
Ambient
Chemical Labeling
NHS Ester
Powder
DMF, DMSO
No
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.