missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Prestained Protein MW Marker

Thermo Scientific Prestained Protein Molecular Weight Marker is a mixture of six blue-stained natural proteins (20 to 120 kDa) for use as size standards in protein electrophoresis (SDS-PAGE).

Supplier:  Thermo Scientific™ 26612

 View more versions of this product

Catalog No. PI26612


Explore available promotions

Description

Description

Thermo Scientific™ Prestained Protein Molecular Weight Marker is a mixture of six blue-stained natural proteins (20 to 120kDa) for use as size standards in protein electrophoresis (SDS-PAGE).

Thermo Scientific Prestained Protein Molecular Weight Marker is a mixture of six blue-stained natural proteins (20 to 120 kDa) for use as size standards in protein electrophoresis (SDS-PAGE). The protein standards are supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Component proteins

  • Beta-galactosidase (120 kDa)
  • Bovine serum albumin (85 kDa)
  • Ovalbumin (50 kDa)
  • Cardonic anhydrase (35 kDa)
  • Beta-lactoglobulin (25 kDa)
  • Lysozyme (20 kDa)

Applications

  • Monitoring protein migration during SDS-polyacrylamide gel electrophoresis.
  • Monitoring protein transfer onto membranes after western blotting
  • Sizing of proteins on SDS-polyacrylamide gels and western blots
Specifications

Specifications

Storage Buffer: 62.5 mM Tris · H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% (w/v) SDS, 1 mM DTT, 1.5 mM NaN3 and 33% (v/v) glycerol.

Upon receipt store at -20°C. Product is shipped with an ice pack.
6
Yes
1 color: Blue
120, 85, 50, 35, 25, 20 kDa
2 x 250 μL
Western Blotting, SDS-PAGE
Colorimetric
Protein Molecular Weight (MW) Marker
20 to 120 kDa
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Pierce™
Approved for shipment on Wet or Dry Ice
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.