missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Sodium Deoxycholate Detergent
Water-soluble, ionic detergent used in cell lysis buffers (e.g., RIPA buffer) to disrupt protein interactions for liposome and membrane protein isolation.
$246.48 - $948.52
Specifications
Product Type | Detergent |
---|---|
Content And Storage | Store at -20°C |
Form | Powder |
Reagent Type | Detergent (Pure) |
Description
Thermo Scientific™ Sodium Deoxycholate is an ionic detergent that is especially useful for disrupting and dissociating protein interactions.
Sodium deoxycholate (deoxycholic acid) is a water-soluble, bile-acid, ionic detergent commonly used in protein methods. It is most frequently used as a component of cell lysis buffers (e.g., RIPA buffer) but also has been used for liposome preparation, isolation of membrane proteins and lipids, preventing nonspecific binding in affinity chromatography and a cell culture media supplement.Highlights:
- Popular anionic, bile-acid detergent for many laboratory uses
- Effective in disrupting and dissociating many types of protein interaction
- Can be removed from solution by dialysis
- Useful for elution or regeneration of certain kinds of affinity columns
- High-purity compound with low UV absorptivity
Detergent Properties:
- Alternative Names: Sodium deoxycholic acid; deoxycholate, sodium salt
- Chemical Name: 3, 12-α-Dihydroxy-5β-cholan-24-oic acid, monosodium salt
- Molecular Weight: 414.6
- Detergent Class: Ionic (anionic)
- Aggregation Number: 5 (average)
- Micelle Molecular Weight: 2000 (average)
- Critical Micelle Concentration (CMC): 2 to 6mM (0.083 to 0.249%, w/v)
- Cloud Point: Unknown
- Dialyzable: Yes
Specifications
Detergent | |
Store at -20°C | |
Powder | |
Detergent (Pure) |
For Research Use Only. Not for use in diagnostic procedures.