Learn More
Thermo Scientific™ Pierce™ Trifluoroacetic Acid (TFA), Sequencing grade
Bottles of 99.5%-pure TFA (MW 114.02, CAS 76-05-01) liquid (sold by weight); for HPLC use as an ion-pairing agent in reverse-phase peptide separations.
$282.43 - $1208.27
Specifications
Name Note | Trifluoroacetic Acid (TFA) |
---|---|
Percent Purity | 99.5% (TFA) |
Physical Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PI28904
|
Thermo Scientific™
LSG28904 |
10 x 1 mL |
Pack of 10 Each for $282.43
|
|
|||||
PI28902
|
Thermo Scientific™
28902 |
10 x 1 g |
Each for $351.00
|
|
|||||
PI28903
|
Thermo Scientific™
28903 |
100 g |
Pack for $511.99
|
|
|||||
PI28901
|
Thermo Scientific™
28901 |
500 mL |
Pack for $1,208.27
|
|
|||||
Description
Thermo Scientific™ Pierce Trifluoroacetic Acid (TFA) is manufactured and tested to meet strict specifications that ensure superior performance for use as an ion-pairing agent in reverse-phase peptide separations.
For complex peptide separations, the key to success can be to vary selectivity. Varying mobile phase composition on the same column can change selectivity enough to resolve peptide that would otherwise overlap. The TFA concentration is usually specified as 0.1% for reverse-phase HPLC of peptides. For reproducible separations from run-to-run or from lab-to-lab, it is essential to make concentrations the same.
Highlights:
- Purity: >99.5% TFA and exceptional clarity allows sensitive, nondestructive peptide detection at low UV wavelengths
- High-performance packaging: Packaged under nitrogen in amber glass with protective TFE-lined fluorocarbon caps to ensure TFA integrity
- Choice of formats for convenience: 1 mL ampules can prepare 1 L of 0.1% v/v TFA solution for the mobile phase in reverse-phase chromatography in moments
- CAS: 76-05-1
Storage Information
- Store in original container protected from direct sunlight in a dry, cool and well-ventilated area
- Should be stored from 20° to 25°C
Order Info
X in the ordering table indicates that hazardous shipping charges apply.
Specifications
Trifluoroacetic Acid (TFA) | |
99.5% (TFA) | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.