missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Water, LC-MS Grade

Thermo Scientific™ Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC

Supplier:  Thermo Scientific™ T51140K2

 View more versions of this product

Catalog No. PI51140


Explore available promotions

Description

Description

Thermo Scientific™ Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC.

An ultrapure, LC/MS-grade preparation with low UV-absorptivity that makes it suitable and reliable for use in HPLC and mass spectrometry applications.

Thermo Scientific Water, LC/MS Grade is specially purified by a proprietary method and treated to ensure lot-to-lot consistency with a low UV absorbance to provide you with most sensitive detection across all wavelengths. Pierce Water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmoshere with Teflon TFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • Purity and quality validated with 30 tests for use in LC/MS applications
  • 0.2-micron filtered, packed in solvent-rinsed amber glass bottles
  • Sealed under nitrogen atmosphere to eliminate absorption of unknown atmospheric gases

Product Details:

General properties of water

  • Molecular formula: H2
  • Molecular Weight: 18.01
  • CAS number: 7732-18-5

Water, LC/MS Grade is specifically purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and prolong the life of your equipment.

Specifications

Specifications

6 to 8
32°F
LC-MS
Liquid
Clear, Colorless
212°F
1 L
Pierce™
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.