3B Scientific™ Pig Embryology Slide Set
Sus scrofa
Supplier:  3B Scientific™ W13058
Catalog No. S95068L
Explore available promotions
Description
Description
10 microscope slides:
  • 1(g). 4-6 mm, t.s.
  • 2(g). 7-9 mm, sagittal l.s.
  • 3(f). 11-12 mm, t.s. through head
  • 4(f). 11-12 mm, t.s. through abdomen
  • 5(f). 15 mm, t.s. through head
  • 6(f). 15 mm, t.s. through thorax
  • 7(f). 15 mm, t.s. through abdomen
  • 8(g). 15 mm, sagittal l.s.
  • 9(g). 20-25 mm, sagittal l.s.
  • 10(g). 20-25 mm, frontal l.s.
Specifications
Specifications
Pig Embryo
10