New Pig PIG™ Green Ham-O™ Absorbent Mat Roll
Brings the fun to leak, drip and spill cleanup. Ham-O layer camouflages liquids and grime while it absorbs; tear off only what you need.
Supplier:  New Pig MAT112
Catalog No. 19003220
Explore available promotions
Description
Description
  • PIG-Printed wide medium weight rolls are ideal for most messes, absorbing everyday spills, leaks and overspray
  • Absorbs most common industrial liquids - oils, coolants, solvents, water and more
  • Scuff-resistant layer stands up to foot traffic in aisles and walkways
  • Won't rip, tear or fray even when saturated; liquids absorb quickly for faster, easier cleanups and safer workplaces
  • Use anywhere - Easy-tear perforations let you tear off small or large pieces for less waste
Specifications
Specifications
Green Camouflage
47.4 gal.
Absorbing and Hiding Spills, Medium Foot Traffic
200 ft.
32 in.
36 lb.
Flame resistant - will melt and self-extinguish, 25% or more Recycled Content
179.4 L
1 Roll/Bag
60.96 m
81.28 cm
16.33 kg