New Pig PIG™ Oil-Only Absorbent Mat Pad
Lasts 2 times longer than ordinary mats for fewer change-outs, while only absorbing oil and fuels, NOT water.
Supplier: New Pig MAT403
Description
- Eight layers of 100% polypropylene are thermally bonded to make PIG Mat the strongest mat on the market; won't rip, tear or fray even when saturated
- Absorbs oil and fuels, NOT water
- White color tells you it's only for oils and fuels; draws attention to machine leaks and clearly shows saturation level during spill response
- Floats to clean up spills on water
- Easy-tear perforations let you take only what you need so you use less mat and save money
Specifications
White | |
83.27 L | |
Absorbing Oil Only | |
20 in. | |
15 in. | |
17 lb. |
22 gal. | |
Polypropylene | |
100 Pads | |
50.8 cm | |
38.1 cm | |
7.71 kg |