missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pimelic Acid, 98%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor P209725KGBL
Specifications
| Pimelic Acid | |
| 1 | |
| Amber Glass Bottle | |
| 160.17 g/mol | |
| Reagent |
| 111-16-0 | |
| White | |
| 25 kg | |
| 0.98 | |
| Crystals or Flakes |
Chemical Identifiers
| 111-16-0 | |
| 160.17 g/mol |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.