missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Piperazine-2-carboxylic acid dihydrochloride, 98%, Thermo Scientific™

Chemical Identifiers

CAS 3022-15-9
Molecular Formula C5H10N2O2·2ClH
Molecular Weight (g/mol) 203.07
MDL Number MFCD00044803
InChI Key WNSDZBQLMGKPQS-UHFFFAOYSA-N
Synonym piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid dihydrochloride, piperazine-2-carboxylic acid 2hcl, +/--piperazine-2-carboxylic acid 2hcl, piperazin-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, +/--2-piperazinecarboxylic acid dihydrochloride, +/--piperazine-2-carboxylic acid dihydrochloride, przca-2hcl, r-+-piperazine-2-carboxylic acid dihydrochloride
PubChem CID 2723757
IUPAC Name piperazine-2-carboxylic acid;dihydrochloride
SMILES C1CNC(CN1)C(=O)O.Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC133660100
SDS View Documents
Thermo Scientific Chemicals
133660100
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC133660500
SDS View Documents
Thermo Scientific Chemicals
133660500
50g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3022-15-9
203.07
WNSDZBQLMGKPQS-UHFFFAOYSA-N
2723757
C1CNC(CN1)C(=O)O.Cl.Cl
C5H10N2O2·2ClH
MFCD00044803
piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid dihydrochloride, piperazine-2-carboxylic acid 2hcl, +/--piperazine-2-carboxylic acid 2hcl, piperazin-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, +/--2-piperazinecarboxylic acid dihydrochloride, +/--piperazine-2-carboxylic acid dihydrochloride, przca-2hcl, r-+-piperazine-2-carboxylic acid dihydrochloride
piperazine-2-carboxylic acid;dihydrochloride

Specifications

3022-15-9
100.0
97.5 to 102.5% (Argentometry)
C5H10N2O2·2ClH
10g
WNSDZBQLMGKPQS-UHFFFAOYSA-N
piperazine-2-carboxylic acid;dihydrochloride
2723757
98%
97.5
Authentic
Glass bottle
MFCD00044803
piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid dihydrochloride, piperazine-2-carboxylic acid 2hcl, +/--piperazine-2-carboxylic acid 2hcl, piperazin-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, +/--2-piperazinecarboxylic acid dihydrochloride, +/--piperazine-2-carboxylic acid dihydrochloride, przca-2hcl, r-+-piperazine-2-carboxylic acid dihydrochloride
C1CNC(CN1)C(=O)O.Cl.Cl
203.07
203.07
Piperazine-2-carboxylic acid dihydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only