missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperazine Dihydriodide 98.0+%, TCI America™
$111.84 - $331.81
Chemical Identifiers
| CAS | 58464-47-4 |
|---|---|
| Molecular Formula | C4H12I2N2 |
| Molecular Weight (g/mol) | 341.96 |
| MDL Number | MFCD00054377 |
| InChI Key | QZCGFUVVXNFSLE-UHFFFAOYSA-N |
| Synonym | Piperazinium Diiodide, Piperazinium Iodide |
| PubChem CID | 20068687 |
| IUPAC Name | piperazine dihydroiodide |
| SMILES | I.I.C1CNCCN1 |
Chemical Identifiers
| 58464-47-4 | |
| 341.96 | |
| QZCGFUVVXNFSLE-UHFFFAOYSA-N | |
| 20068687 | |
| I.I.C1CNCCN1 |
| C4H12I2N2 | |
| MFCD00054377 | |
| Piperazinium Diiodide, Piperazinium Iodide | |
| piperazine dihydroiodide |
Specifications
| 58464-47-4 | |
| C4H12I2N2 | |
| 1 g | |
| QZCGFUVVXNFSLE-UHFFFAOYSA-N | |
| piperazine dihydroiodide | |
| 20068687 | |
| ≥98.0% (N,T) | |
| Piperazine Dihydriodide |
| 250°C | |
| MFCD00054377 | |
| Piperazinium Diiodide, Piperazinium Iodide | |
| I.I.C1CNCCN1 | |
| 341.96 | |
| 341.96 | |
| Crystalline Powder |
Safety and Handling
TSCA : No