missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperazine Dihydrochloride Monohydrate 98.0+%, TCI America™
$61.51 - $301.98
Chemical Identifiers
| CAS | 142-64-3 |
|---|---|
| Molecular Formula | C4H12Cl2N2 |
| Molecular Weight (g/mol) | 159.054 |
| MDL Number | MFCD00012754 |
| InChI Key | CVVIJWRCGSYCMB-UHFFFAOYSA-N |
| Synonym | piperazine dihydrochloride, dowzene dhc, piperazine, dihydrochloride, dihydro pip wormer, caswell no. 668, piperazine wormer premix, unii-17vu4z4w88, diethylenediamine dihydrochloride, piperazine hcl, epa pesticide chemical code 067401 |
| PubChem CID | 8893 |
| IUPAC Name | piperazine;dihydrochloride |
| SMILES | C1CNCCN1.Cl.Cl |
Chemical Identifiers
| 142-64-3 | |
| 159.054 | |
| CVVIJWRCGSYCMB-UHFFFAOYSA-N | |
| 8893 | |
| C1CNCCN1.Cl.Cl |
| C4H12Cl2N2 | |
| MFCD00012754 | |
| piperazine dihydrochloride, dowzene dhc, piperazine, dihydrochloride, dihydro pip wormer, caswell no. 668, piperazine wormer premix, unii-17vu4z4w88, diethylenediamine dihydrochloride, piperazine hcl, epa pesticide chemical code 067401 | |
| piperazine;dihydrochloride |
Specifications
| 142-64-3 | |
| White | |
| MFCD00012754 | |
| piperazine dihydrochloride, dowzene dhc, piperazine, dihydrochloride, dihydro pip wormer, caswell no. 668, piperazine wormer premix, unii-17vu4z4w88, diethylenediamine dihydrochloride, piperazine hcl, epa pesticide chemical code 067401 | |
| C1CNCCN1.Cl.Cl | |
| 159.054 | |
| 159.05 | |
| Crystalline Powder |
| 320°C | |
| C4H12Cl2N2 | |
| 25 g | |
| CVVIJWRCGSYCMB-UHFFFAOYSA-N | |
| piperazine;dihydrochloride | |
| 8893 | |
| ≥98.0% (N) | |
| Piperazine Dihydrochloride Monohydrate |
Safety and Handling
RTECSNumber : TL4025000
TSCA : Yes